Skip to Content
Merck
All Photos(1)

Key Documents

GF95357526

Carbon - Vitreous

foil, 100x100mm, thickness 1.0mm, glassy carbon

Synonym(s):

Carbon - Vitreous, VC000500

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
CAS Number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23

form

foil

manufacturer/tradename

Goodfellow 953-575-26

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antoine P Pagé et al.
PloS one, 10(7), e0132062-e0132062 (2015-07-15)
The objectives of this study were to uncover Salix purpurea-microbe xenobiotic degradation systems that could be harnessed in rhizoremediation, and to identify microorganisms that are likely involved in these partnerships. To do so, we tested S. purpurea's ability to stimulate
Svenja T Lohner et al.
The ISME journal, 8(8), 1673-1681 (2014-05-23)
Direct, shuttle-free uptake of extracellular, cathode-derived electrons has been postulated as a novel mechanism of electron metabolism in some prokaryotes that may also be involved in syntrophic electron transport between two microorganisms. Experimental proof for direct uptake of cathodic electrons
Catharina Vendl et al.
The Journal of experimental biology, 218(Pt 21), 3425-3434 (2015-11-06)
Fundamental differences in methane (CH4) production between macropods (kangaroos) and ruminants have been suggested and linked to differences in the composition of the forestomach microbiome. Using six western grey kangaroos (Macropus fuliginosus) and four red kangaroos (Macropus rufus), we measured
Matthias Egger et al.
Environmental science & technology, 49(1), 277-283 (2014-11-21)
Methane is a powerful greenhouse gas and its biological conversion in marine sediments, largely controlled by anaerobic oxidation of methane (AOM), is a crucial part of the global carbon cycle. However, little is known about the role of iron oxides
A Romero-Perez et al.
Journal of animal science, 93(4), 1780-1791 (2015-05-29)
The objective was to evaluate whether long-term addition of 3-nitrooxypropanol (NOP) to a beef cattle diet results in a sustained reduction in enteric CH4 emissions in beef cattle. Eight ruminally cannulated heifers (637 ± 16.2 kg BW) were used in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service