Key Documents
ESYNTH019
Sigma-Aldrich
SynLectro Glassy Carbon Electrode
size 3 mm × 20 mm × 60 mm
Synonym(s):
Glassy Carbon SynLectro Electrode
About This Item
UNSPSC Code:
81141502
NACRES:
NB.61
Pricing and availability is not currently available.
size
3 mm × 20 mm × 60 mm
General description
SynLectro Glassy Carbon Electrode is a 3x20x60 mm electrode designed for use with the SynLectro Electrolysis Platform for synthetic electrochemistry.
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. Is there any kind of documentation on the elemental and/or structural analysis of the electrode? I.e. what percentage is amorphous carbon vs graphite?
    1 answer
    1. There is no documentation on the elemental and/or structural analysis of the electrode, including the percentages of amorphous carbon versus graphite. This product is made from glassy carbon, which is a non-graphitizing carbon material.
      Helpful?
  2. Is it possible to cut the electrode ESYNTH019-1EA into smaller pieces, for example, cutting it in half to make 3mm x 10mm x 30mm from the original 3 mm × 20 mm × 60 mm? If so, what is the best method for doing so - diamond scribe or some other method?
    1 answer
    1. In the past, diamond scribers have been used for this purpose. However, this method requires quickly pressing an edge against it on the reverse side. If the customer has access to a mechanical workshop, a diamond saw is recommended. It should be noted that glassy carbon becomes harder with higher pressure. Therefore, when using a diamond saw, it is advisable to saw slowly and not apply excessive pressure.
      Helpful?