Skip to Content
Merck
All Photos(2)

Key Documents

GF88214542

Gold

insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%

Synonym(s):

Gold, AU005831

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99%

form

wire

manufacturer/tradename

Goodfellow 882-145-42

resistivity

2.05 μΩ-cm, 0°C

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Sun et al.
Journal of nanoscience and nanotechnology, 14(6), 4078-4081 (2014-04-18)
Colloidal gold is extensively used for molecular sensing because that the surface plasmon resonance (SPR) bands are affected by changes in the dielectric properties in the close vicinity of these structures due to the binding of ligands to the corresponding
Celina Yang et al.
Journal of nanoscience and nanotechnology, 14(7), 4813-4819 (2014-04-25)
The applications of nanoparticles (NPs) for improved therapeutics are at the forefront of cancer nanotechnology. Gold nanoparticles (GNPs) have been extensively used due to their ability to act as both an anticancer drug carrier in chemotherapy and as a dose
Kaili Niu et al.
Journal of nanoscience and nanotechnology, 14(7), 5151-5156 (2014-04-25)
An immunochromatographic test strip using gold nanoparticles-staphylococcus aureus monoclonal antibody conjugates was developed for the rapid and convenient detection of staphylococcus aureus based on a double-antibody sandwich format. The detection limit and the detection rate of this test strip is
Lin Luo et al.
Journal of nanoscience and nanotechnology, 14(6), 4036-4042 (2014-04-18)
A novel immunosensor of human immune globulin (IgG) was fabricated based on the fluorescence transfer between luminescent semiconductor quantum dots (QDs) and gold nanoparticles (AuNPs). AuNPs and CdSe/ZnS QDs were respectively labeled with immune reaction pair:IgG and goat anti-human immunoglobulin
Maria Jose Santos-Martinez et al.
Journal of biomedical nanotechnology, 10(6), 1004-1015 (2014-04-23)
The increasing use of gold nanoparticles in medical diagnosis and treatment has raised the concern over their blood compatibility. The interactions of nanoparticles with blood components may lead to platelet aggregation and endothelial dysfunction. Therefore, medical applications of gold nanoparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service