Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold coated glass slide, 99.999% (Au), layer thickness 100 Å
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold coated glass cover slip, L × W × thickness 22 mm × 22 mm × 130-170 μm, square, 99.999% (Au), layer thickness 100 Å
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Gold, insulated wire, 5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Gold, microfoil, disks, 10mm, thinness 0.75μm, specific density 1449μg/cm2, permanent mylar 3.5μm support, 99.99+%
Gold, insulated wire, 1m, conductor diameter 0.125mm, insulation thickness 0.016mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, insulated wire, 2m, conductor diameter 0.05mm, insulation thickness 0.009mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, insulated wire, 0.5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Gold, tube, 200mm, outside diameter 1.0mm, inside diameter 0.7mm, wall thickness 0.15mm, as drawn, 99.95%
Gold, insulated wire, 1m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Gold, tube, 50mm, outside diameter 1.60mm, inside diameter 0.6mm, wall thickness 0.5mm, as drawn, 99.95%
Gold, tube, 200mm, outside diameter 1.60mm, inside diameter 0.6mm, wall thickness 0.5mm, as drawn, 99.95%
Gold, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.009mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, rod, 10mm, diameter 5.0mm, as drawn, 99.95%
Gold, rod, 50mm, diameter 1.6mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 2.0mm, inside diameter 1.8mm, wall thickness 0.1mm, as drawn, 99.95%
Gold, rod, 25mm, diameter 2.0mm, as drawn, 99.95%
Gold, sphere, 20pcs, diameter 0.5mm, 99.99%
Gold, tube, 100mm, outside diameter 2.0mm, inside diameter 1.7mm, wall thickness 0.15mm, as drawn, 99.95%
Gold, tube, 50mm, outside diameter 2.0mm, inside diameter 1.7mm, wall thickness 0.15mm, as drawn, 99.95%