Skip to Content
Merck

GF82934703

Iridium

foil, thickness 0.1 mm, size 25 × 25 mm, purity 99.9%

Synonym(s):

IR000209

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ir
CAS Number:
Molecular Weight:
192.22
MDL number:
UNSPSC Code:
12141720
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

Assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 829-347-03

resistivity

4.71 μΩ-cm

size

25 × 25 mm

thickness

0.1 mm

bp

4130 °C (lit.)

mp

2450 °C (lit.)

density

22.65 g/cm3 (lit.)

SMILES string

[Ir]

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
E8909SAB2500767HPA004729
specific activity

≥2,000 unit/μg protein

specific activity

≥1000 units/μg protein

specific activity

-

specific activity

-

form

solution

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous glycerol solution

recombinant

expressed in Sf9 cells

recombinant

expressed in Sf9 cells

recombinant

-

recombinant

-

mol wt

126 kDa

mol wt

-

mol wt

-

mol wt

-

UniProt accession no.

O76074

UniProt accession no.

-

UniProt accession no.

O76074

UniProt accession no.

O76074

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service