GF50243284
Bismuth
foil, not light tested, 150x150mm, thickness 0.05mm, permanent polyester support, 99.97%
Synonym(s):
Bismuth, BI000190
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
<0.1 mmHg ( 20 °C)
Assay
99.97%
form
foil
manufacturer/tradename
Goodfellow 502-432-84
resistivity
129 μΩ-cm, 20°C
size × thickness
150 x 150 mm × 0.05 mm
bp
1560 °C (lit.)
mp
271 °C (lit.)
density
9.8 g/mL at 25 °C (lit.)
SMILES string
[Bi]
InChI
1S/Bi
InChI key
JCXGWMGPZLAOME-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Javier P Gisbert et al.
Helicobacter, 10(3), 157-171 (2005-05-21)
To systematically review the Helicobacter pylori eradication efficacy with ranitidine bismuth citrate (RBC) and two antibiotics, and to conduct a meta-analysis of randomized clinical trials comparing the efficacy of proton pump inhibitor (PPI) vs. RBC with two antibiotics for 1
Montserrat Filella
Journal of environmental monitoring : JEM, 12(1), 90-109 (2010-01-19)
Like all elements of the periodic table, bismuth is ubiquitously distributed throughout the environment as a result of natural processes and human activities. It is present as Bi(III) in environmental, biological and geochemical samples. Although bismuth and its compounds are
Emma Aneheim et al.
Scientific reports, 5, 12025-12025 (2015-07-15)
To facilitate multicentre clinical studies on targeted alpha therapy, it is necessary to develop an automated, on-site procedure for conjugating rare, short-lived, alpha-emitting radionuclides to biomolecules. Astatine-211 is one of the few alpha-emitting nuclides with appropriate chemical and physical properties
Bismuth in medicine.
Hongzhe Sun et al.
Metal ions in biological systems, 41, 333-378 (2004-06-23)
Jorge A R Salvador et al.
Molecules (Basel, Switzerland), 16(4), 2884-2913 (2011-04-06)
Steroid and terpene chemistry still have a great impact on medicinal chemistry. Therefore, the development of new reactions or "greener" processes in this field is a contemporaneous issue. In this review, the use of bismuth(III) salts, as "ecofriendly" reagents/catalysts, on
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service