Skip to Content
Merck
Bismuth foil, not light tested, 150x150mm, thickness 0.025mm, permanent polyester support, 99.97%

Bismuth

Bismuth foil, not light tested, 150x150mm, thickness 0.025mm, permanent polyester support, 99.97%
Synonym(s):
Bismuth
Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Element
Quality Segment
Showing 1-20 of 103
Page 1 of 6
Compare
Product Number
Description
Pricing
Page 1 of 6