GF23358078
Titanium/Aluminum/Vanadium
powder, Ti 90%/Al 6%/V 4%, max. particle size 200 micron, weight 200 g, min. particle size 150 micron
Synonym(s):
TI016015
About This Item
Recommended Products
form
powder
manufacturer/tradename
Goodfellow 233-580-78
weight
200 g
max. part. size
200 μm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service