Skip to Content
Merck
All Photos(2)

Key Documents

GF12515891

Bismuth

foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.97%

Synonym(s):

Bismuth, BI000140

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23

Pricing and availability is not currently available.

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.97%

form

foil

manufacturer/tradename

Goodfellow 125-158-91

resistivity

129 μΩ-cm, 20°C

size × thickness

25 x 25 mm × 0.015 mm

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
171450610006121044662
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

API family

vinorelbine

API family

vinorelbine

API family

acebutolol

API family

atovaquone

storage temp.

−20°C

storage temp.

−70°C

storage temp.

-

storage temp.

2-8°C

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Javier P Gisbert et al.
    Helicobacter, 10(3), 157-171 (2005-05-21)
    To systematically review the Helicobacter pylori eradication efficacy with ranitidine bismuth citrate (RBC) and two antibiotics, and to conduct a meta-analysis of randomized clinical trials comparing the efficacy of proton pump inhibitor (PPI) vs. RBC with two antibiotics for 1
    Montserrat Filella
    Journal of environmental monitoring : JEM, 12(1), 90-109 (2010-01-19)
    Like all elements of the periodic table, bismuth is ubiquitously distributed throughout the environment as a result of natural processes and human activities. It is present as Bi(III) in environmental, biological and geochemical samples. Although bismuth and its compounds are
    Emma Aneheim et al.
    Scientific reports, 5, 12025-12025 (2015-07-15)
    To facilitate multicentre clinical studies on targeted alpha therapy, it is necessary to develop an automated, on-site procedure for conjugating rare, short-lived, alpha-emitting radionuclides to biomolecules. Astatine-211 is one of the few alpha-emitting nuclides with appropriate chemical and physical properties
    Jorge A R Salvador et al.
    Molecules (Basel, Switzerland), 16(4), 2884-2913 (2011-04-06)
    Steroid and terpene chemistry still have a great impact on medicinal chemistry. Therefore, the development of new reactions or "greener" processes in this field is a contemporaneous issue. In this review, the use of bismuth(III) salts, as "ecofriendly" reagents/catalysts, on
    Syntheses of biodegradable and biocompatible polymers by means of bismuth catalysts.
    Hans R Kricheldorf
    Chemical reviews, 109(11), 5579-5594 (2009-09-26)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service