Skip to Content
Merck
All Photos(2)

Key Documents

G307

Sigma-Aldrich

Galvinoxyl, free radical

Synonym(s):

2,6-Di-tert-butyl-α-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H41O2
CAS Number:
Molecular Weight:
421.63
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

mp

158-159 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)c1cc(\C=C2\C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)cc(c1[O])C(C)(C)C

InChI

1S/C29H41O2/c1-26(2,3)20-14-18(15-21(24(20)30)27(4,5)6)13-19-16-22(28(7,8)9)25(31)23(17-19)29(10,11)12/h13-17H,1-12H3

InChI key

GNZDAXRYGVFYPU-UHFFFAOYSA-N

Application

Free radical and free-radical scavenger.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuichi Shimakawa et al.
Lipids, 38(3), 225-231 (2003-06-06)
Nitroxyl radicals are known to act as radical scavenging antioxidants. In the present study, a lipophilic nitroxyl radical, cyclohexane-1-spiro-2'-(4'-oxyimidazolidine-1'-oxyl)-5'-spiro-1"-cyclohexane (nitroxyl radical I) was synthesized and its antioxidant capacity was assessed in comparison with a hydrophilic nitroxyl radical, 4-hydroxy-2,2,6,6-tetra-methylpiperidine-N-oxyl (Tempol). Both
Lan-Fen Wang et al.
Bioorganic chemistry, 33(2), 108-115 (2005-03-25)
To improve the radical-scavenging activity of catechin, a planar catechin analogue was designed and synthesized by Fukuhara [J. Am. Chem. Soc. 124 (2002) 5952]. Although the planar catechin is less active than quercetin, it is much more active than catechin
Y Okada et al.
Redox report : communications in free radical research, 6(2), 117-122 (2001-07-14)
The antioxidant activity of capsaicin (CAP) was measured in the oxidation of methyl linoleate (ML) in homogeneous solution, of ML micelles in aqueous dispersions and also of soybean phosphatidylcholine liposomal membrane, and was compared to that of alpha-tocopherol (alpha-TOH) which
Galvinoxyl method for standardizing electron and proton donation activity.
H Shi et al.
Methods in enzymology, 335, 157-166 (2001-06-13)
Jian-Ying Feng et al.
Journal of agricultural and food chemistry, 57(22), 11041-11046 (2009-09-10)
The aim of this work is to clarify the antioxidant abilities of phenolic and enolic hydroxyl groups in curcumin. 1,7-bis(4-benzyloxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione (BEC), 1,7-bis(4-hydroxy-3-methoxyphenyl)heptane-3,5-diol (OHC), 1,7-bis(4-hydroxy-3-methoxyphenyl)heptane-3,5-dione (THC), and 1,7-bis(3,4-dihydroxyphenyl)-1,6-heptadiene-3,5-dione (BDC) are synthesized to determine the antioxidant activities by using antiradical assays against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service