Skip to Content
Merck
All Photos(1)

Key Documents

D206806

Sigma-Aldrich

1,1-Diphenylethylene

97%

Synonym(s):

α,α-Diphenylethylene, 1,1-Diphenylethene, 1,1′-Ethenylidenebis[benzene], 1-Phenylethenylbenzene, Rhodium acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2C=CH2
CAS Number:
Molecular Weight:
180.25
Beilstein:
1099062
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.608 (lit.)

bp

270-271 °C (lit.)

mp

6 °C (lit.)

density

1.021 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=C(c1ccccc1)c2ccccc2

InChI

1S/C14H12/c1-12(13-8-4-2-5-9-13)14-10-6-3-7-11-14/h2-11H,1H2

InChI key

ZMYIIHDQURVDRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongru Li et al.
Journal of fluorescence, 21(4), 1721-1728 (2011-03-04)
This article presents a comprehensive theoretical investigation of excited state intramolecular proton transfer (ESIPT) for some newly-designed diphenylethylene derivatives containing 2-(2-hydroxy-phenyl)-benzotriazole moiety with various substituted groups. The calculation shows the structural parameters and Mulliken charges of phototautomers enol (E) and
Emmanouil D Tsochatzis et al.
Analytica chimica acta, 1130, 49-59 (2020-09-08)
A simple, fast, sensitive and reliable method was developed for the simultaneous determination of 13 food contact materials (FCM) regulated substances and non-intentionally added substances (NIAS) migrating into official food simulants. The method has been optimized to quantify the monomers
Debajyoti Ray et al.
Environmental science & technology, 45(23), 10061-10067 (2011-11-11)
We measured the kinetics of ozonation reaction of 1,1-diphenylethylene (DPE) in artificial snow, produced by shock freezing of DPE aqueous solutions sprayed into liquid nitrogen. It was demonstrated that most of the reactant molecules are in direct (productive) contact with
Michael J Minkler et al.
Magnetic resonance in chemistry : MRC, 58(12), 1168-1176 (2020-04-09)
Low-cost, high-accuracy characterization of polymeric materials is critical for satisfying societal demand for high-quality materials with ultra-specific requirements. Low-field nuclear magnetic resonance (NMR) spectroscopy presents an opportunity to replace costlier or destructive methods while utilizing nondeuterated solvents. Many factors play
E Bignon et al.
Biochemical and biophysical research communications, 166(3), 1471-1478 (1990-02-14)
Protein kinase C (PKC) I (gamma), II (beta) and III (alpha) subspecies are all activated by 1,1-di-(p-hydroxyphenyl)ethylene derivatives (DPE) at micromolar concentrations. This PKC activation depends on the presence of both Ca2+ and phosphatidylserine (PS) but does not require diacylglycerol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service