Skip to Content
Merck
All Photos(1)

Documents

F9108

Sigma-Aldrich

5-Fluoroindole

98%

Synonym(s):

NSC 88613

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6FN
CAS Number:
Molecular Weight:
135.14
Beilstein:
112350
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

45-48 °C (lit.)

SMILES string

Fc1ccc2[nH]ccc2c1

InChI

1S/C8H6FN/c9-7-1-2-8-6(5-7)3-4-10-8/h1-5,10H

InChI key

ODFFPRGJZRXNHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Reactant for preparation of 5-HT6 receptor ligands
  • Reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Reactant for preparation of antitumor agents
  • Reactant for preparation of antibacterial agents
  • Reactant for preparation of immunosuppressive agents
  • Reactant for preparation of Sodium-Dependent Glucose Co-transporter 2 (SGLT2) Inhibitors for the Management of Hyperglycemia in Diabetes
  • Reactant for preparation of Myeloperoxidase Inhibitors
  • Reactant for preparation of Potent Selective Serotonin Reuptake Inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steffen P Graether et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 178(1), 65-71 (2005-10-04)
We show that it is feasible to use a minicoil for solid-state 19F 1H NMR experiments that has short pulse widths, good RF homogeneity, and excellent signal-to-noise for small samples while using low power amplifiers typical to liquid-state NMR. The
Peter B Crowley et al.
Chemical communications (Cambridge, England), 48(86), 10681-10683 (2012-09-25)
Fluorine-containing amino acids are valuable probes for the biophysical characterization of proteins. Current methods for (19)F-labeled protein production involve time-consuming genetic manipulation, compromised expression systems and expensive reagents. We show that Escherichia coli BL21, the workhorse of protein production, can
Nicholas A Magnus et al.
Organic letters, 12(16), 3700-3703 (2010-08-14)
A practical synthesis of the glycogen synthase kinase-3 (GSK3) inhibitor bisarylmaleimide 1 has been accomplished employing Pictet-Spengler methodology to access the indole 7-position in preparing the benzodiazepine tricyclic fragment. A seven-step linear sequence that starts with commercially available 5-fluoroindole 7
A L Palombella et al.
Plant physiology, 117(2), 455-464 (1998-06-25)
We report the isolation of a Chlamydomonas reinhardtii cDNA that encodes the beta-subunit of tryptophan synthase (TSB). This cDNA was cloned by functional complementation of a trp-operon-deleted strain of Escherichia coli. Hybridization analysis indicated that the gene exists in a
Jia Bai et al.
Biochimica et biophysica acta. Biomembranes, 1862(5), 183194-183194 (2020-01-19)
Microsomal cytochrome b5 (cytb5) is a membrane-bound protein capable of donating the second electron to cytochrome P450s (cytP450s) in the cytP450s monooxygenase reactions. Recent studies have demonstrated the importance of the transmembrane domain of cytb5 in the interaction with cytP450

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service