Documents
ESYNTH001
Sigma-Aldrich
SynLectro 50 mL starter kit
Synonym(s):
SynLectro 50 mL Starter Set
About This Item
UNSPSC Code:
81141502
NACRES:
NB.61
General description
SynLectro 50 mL Starter Kit is the perfect way to get your electrochemical reaction started.
This kit contains:
  • 50 mL glass cell
  • PTFE stopper and sleeve
  • 2 electrode holders
  • 2 graphite electrodes
  • stainless steel electrode
  • nickel electrode
  • lead electrode
  • zinc electrode
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany