Skip to Content
Merck
All Photos(1)

Documents

E3708

Sigma-Aldrich

Ethanethiol

97%

Synonym(s):

Ethyl mercaptan, Mercaptan C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SH
CAS Number:
Molecular Weight:
62.13
Beilstein:
773638
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.1 (vs air)

Quality Level

vapor pressure

8.51 psi ( 20 °C)

Assay

97%

form

liquid

autoignition temp.

570 °F

expl. lim.

18.2 %

refractive index

n20/D 1.4306 (lit.)

bp

35 °C (lit.)

density

0.839 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCS

InChI

1S/C2H6S/c1-2-3/h3H,2H2,1H3

InChI key

DNJIEGIFACGWOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethanethiol can be used as a reactant for the synthesis of:
  • Functionalized oxazolidinones by conjugated nucleophilic addition- electrophilic amination reaction.
  • Ethyl phenyl sulfide by C-S coupling with iodobenzene.
  • Chain transfer agent in the RAFT polymerization of N-isopropylacrylamide.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-49.0 °F - closed cup

Flash Point(C)

-45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct synthesis of thermally responsive DMA/NIPAM diblock and DMA/NIPAM/DMA triblock copolymers via aqueous, room temperature RAFT polymerization
Convertine AJ, et al.
Macromolecules, 39(5), 1724-1730 (2006)
Asymmetric multicomponent domino reactions and highly enantioselective conjugated addition of thiols to ?, ?-unsaturated aldehydes
Marigo M, et al
Journal of the American Chemical Society, 127(45), 15710-15711 (2005)
Efficient CuO-Nanoparticle-Catalyzed C-S Cross-Coupling of Thiols with Iodobenzene
Rout L, et al.
Angewandte Chemie (International Edition in English), 119(29), 5679-5682 (2007)
Jiangyao Chen et al.
Journal of hazardous materials, 190(1-3), 416-423 (2011-04-20)
A series of adsorptive photocatalysts, combined titania-montmorillonite-silica were synthesized. The resultant photocatalysts consisted of more and more spherically agglomerated TiO(2) particles with increasing of TiO(2) content, and anatase was the only crystalline phase with nano-scale TiO(2) particles. With increasing of
R Hanczkó et al.
Journal of chromatography. A, 1163(1-2), 25-42 (2007-07-04)
The main aims of this work were (a) to present the characteristics and stability of the o-phthalaldehyde (OPA)-ethanethiol (ET) derivatives of 22 amino acids, including the believed-to-be less stable OPA derivatives providing glycine, gamma-aminobutyric acid, beta-alanine, histidine, ornithine, lysine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service