Skip to Content
Merck
All Photos(1)

Key Documents

T32808

Sigma-Aldrich

Thiophenol

97%

Synonym(s):

Benzenethiol, Phenyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SH
CAS Number:
Molecular Weight:
110.18
Beilstein:
506523
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

3.8 (vs air)

Quality Level

vapor pressure

1.4 mmHg ( 20 °C)

Assay

97%

refractive index

n20/D 1.588 (lit.)

bp

169 °C (lit.)

mp

−15 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

Sc1ccccc1

InChI

1S/C6H6S/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

RMVRSNDYEFQCLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiophenol is an organosulfur compound used as an intermediate to produce polymers.[1]

Application

Thiophenol can undergo:
  • Asymmetric Michael addition to diisopropyl maleate in the presence of a catalytic amount of cinchonine to form (s)-(−)-diisopropyl phenylthiosuccinate, which can be further converted to (R)-(+)-3,4-epoxy-1-butanol.[2]
  • Regioselective and stereoselective 1,4-addition to alkenylidenecyclopropanes leading to endo- or cis-vinyl sulfide adducts, a key step in the synthesis of karahanaenone.[3]
  • Diastereoselective addition to (S)-N-(α,β-unsaturated carbonyl)-γ-[(trityloxy)methyl]-γ-butyrolactams to form the corresponding β-(phenylthio)carboxylates.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiophenol additions to alkenylidenecyclopropanes. A novel synthesis of karahanaenone.
Cairns P M, et al.
Tetrahedron Letters, 23(13), 1405-1408 (1982)
Palladium-Catalyzed Regioselective Addition of Thiophenol to Conjugated Enynes. Efficient Syntheses of 2-(Phenylsulfinyl) and 2-(Phenylsulfonyl) 1, 3-Dienes.
Baeckvall J-E and Anna E
The Journal of Organic Chemistry, 59(20), 5850-5851 (1994)
Diastereoselective Thiophenol Addition to (S)-N-. alpha.,. beta.-Unsaturated carbonyl-. gamma.-[(trityloxy) methyl]-. gamma.-butyrolactams.
Tomioka K, et al.
The Journal of Organic Chemistry, 60(19), 6188-6190 (1995)
A ratiometric fluorescent probe for selective detection of thiophenol derivatives
Li F, et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 271, 120870-120870 (2022)
Ki-Jeong Kim et al.
Scientific reports, 10(1), 2119-2119 (2020-02-09)
The surface morphology and electronic structure of hexagonal graphene onion rings (HGORs), a modified graphene structure, were investigated to confirm the possibility as an efficient catalyst when compared to graphene. After confirming the formation of HGORs with a smaller width

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service