Skip to Content
Merck
All Photos(1)

Key Documents

E24859

Sigma-Aldrich

Protocatechuic acid ethyl ester

97%

Synonym(s):

Ethyl 3,4-dihydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2C2H5
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

132-134 °C (lit.)

SMILES string

CCOC(=O)c1ccc(O)c(O)c1

InChI

1S/C9H10O4/c1-2-13-9(12)6-3-4-7(10)8(11)5-6/h3-5,10-11H,2H2,1H3

InChI key

KBPUBCVJHFXPOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regina G Daré et al.
International journal of pharmaceutics, 582, 119336-119336 (2020-04-19)
Excessive exposure to solar radiation induces injurious effects on human skin. Our previous study evidenced that protocatechuic acid (P0) and ethyl protocatechuate (P2) act against photodamage and photoaging. The present study aimed to develop solid lipid nanoparticles (SLNs) and nanostructured
S Saika et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 232(8), 499-502 (1994-08-01)
The spreading of epithelium is critical in the healing of corneal wounds. Such epithelial spreading requires the continuous production of protein and glycoprotein. To determine whether collagen production is required for the spreading of corneal epithelium, we studied the effects
Percy W Y Chu et al.
European journal of pharmacology, 633(1-3), 24-32 (2010-02-16)
Tolerance to brain injury involves hypoxia-inducible factor-1 (HIF-1) and its target genes as the key pathway mediating a cascade of events including cell survival, energetics, and angiogenesis. In this study, we established the treatment paradigms for an in vitro model
Shiow Chyn Huang et al.
Journal of agricultural and food chemistry, 51(8), 2380-2383 (2003-04-03)
The antioxidant activity and identification of the antioxidant component of peanut seed testa were investigated. The antioxidant activity of peanut seed testa was studied in the linoleic acid model system by using the ferric thiocyanate method. Among the five organic
Jian Wang et al.
FEBS letters, 529(2-3), 309-312 (2002-10-10)
Ethyl-3,4-dihydroxybenzoate (EDHB) is commonly utilized as a substrate analog and competitive inhibitor of prolyl 4-hydroxylases. These iron-dependent enzymes have received a lot of attention for their involvement in crucial biochemical pathways such as collagen maturation and oxygen sensing. Since EDHB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service