DWKFDF036N
Azlon Polypropylene Volumetric Flask With PPE Stopper
class B, capacity 500 mL , with +/-5mL tolerance
Synonym(s):
FDF036N
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
feature
class B
packaging
pkg of 1 ea
manufacturer/tradename
Azlon FDF036N
capacity
500 mL , with +/-5mL tolerance
Looking for similar products? Visit Product Comparison Guide
General description
With an easy to read calibration line, the AZLON® class B volumetric flask is suitable for accurate measuring and is supplied with a polypropylene stopper. Provide a good alternative to glass.
Application
For the accurate preparation of solutions where glass alternatives would not be suitable
Features and Benefits
Excellent durability and calibrated to ISO 1042
Legal Information
AZLON is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service