Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azlon Polypropylene Volumetric Flask With PPE Stopper
Products
Product Category
flasks (6)
Manufacturer
Capacity (mL)
Glassware Class
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
DWKFDF012N
Azlon Polypropylene Volumetric Flask With PPE Stopper,
class B, capacity 25 mL
Expand
View Pricing
DWKFDF024N
Azlon Polypropylene Volumetric Flask With PPE Stopper,
class B, capacity 100 mL
Expand
View Pricing
DWKFDF038N
Azlon Polypropylene Volumetric Flask With PPE Stopper,
class B, capacity 1000 mL
Expand
View Pricing
DWKFDF036N
Azlon Polypropylene Volumetric Flask With PPE Stopper,
class B, capacity 500 mL , with +/-5mL tolerance
Expand
View Pricing
DWKFDF018N
Azlon Polypropylene Volumetric Flask With PPE Stopper,
class B, capacity 50 mL
Expand
View Pricing
DWKFDF030N
Azlon Polypropylene Volumetric Flask With PPE Stopper,
class B, capacity 250 mL
Expand
View Pricing