Skip to Content
Merck
All Photos(5)

Key Documents

D91551

Sigma-Aldrich

Diethyl carbonate

greener alternative

99%

Synonym(s):

DEC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2CO
CAS Number:
Molecular Weight:
118.13
Beilstein:
956591
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.1 (vs air)

Quality Level

vapor pressure

10 mmHg ( 23.8 °C)
59 mmHg ( 37.8 °C)

Assay

99%

form

liquid

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.384 (lit.)

bp

126-128 °C (lit.)

mp

−43 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O=C(OCC)OCC

InChI

1S/C5H10O3/c1-3-7-5(6)8-4-2/h3-4H2,1-2H3

InChI key

OIFBSDVPJOWBCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is used as a greener reagent. Click here for more information.

Application

Diethyl carbonate (DEC) is generally used in the C-alkoxycarbonylation of enolate anions, aryl and alkyl cyanides. It reacts with 1,2-amino alcohols to yield corresponding 2-oxazolidinones. Additionally, DEC is also used as a component of electrolyte solution in lithium ion batteries.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diastereoselective Aldol Condensation Using a Chiral Oxazolidinone Auxiliary:(2S*, 3S*)-3-Hydroxy-3-phenyl-2-methylpropanoic Acid: Benzenepropanoic acid, β-hydroxy-α-methyl-,[S-(R*, R*)]-
Gage JR and Evans DA
Organic Syntheses, 68, 83-83 (2003)
Thermal stability of alkyl carbonate mixed-solvent electrolytes for lithium ion cells.
Kawamura T, et al.
Journal of Power Sources, 104(2), 260-264 (2002)
Diethyl Carbonate.
Krapcho AP and Gorin DJ
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-10 (2001)
The study of the anodic stability of alkyl carbonate solutions by in situ FTIR spectroscopy, EQCM, NMR and MS.
Moshkovich M, et al.
Journal of Electroanalytical Chemistry, 497(1-2), 84-96 (2001)
Dan Li et al.
Journal of hazardous materials, 161(2-3), 1193-1201 (2008-06-03)
Vapor pressures and flash points for several mixtures of an aviation fuel with dimethyl carbonate (DMC) or diethyl carbonate (DEC) have been measured, respectively, over the entire composition range. Correlation between the experimental vapor pressures and the equilibrium temperatures by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service