Skip to Content
Merck
All Photos(1)

Key Documents

D8022

Sigma-Aldrich

3,5-Dichlorophenyldiazonium tetrafluoroborate

Synonym(s):

3,5-Dichlorobenzenediazonium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2N2 · BF4
CAS Number:
Molecular Weight:
260.81
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

storage temp.

−20°C

SMILES string

F[B-](F)(F)F.Clc1cc(Cl)cc(c1)[N+]#N

InChI

1S/C6H3Cl2N2.BF4/c7-4-1-5(8)3-6(2-4)10-9;2-1(3,4)5/h1-3H;/q+1;-1

InChI key

XSVOZXRITFTBIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Electron transfer chemistry
  • Microwave-accelerated cross-coupling reactions
  • Sandmeyer bromination for preparation of bromobenzene derivatives
  • Catalytic thiocyanation in the presence of copper salts
  • Electrochemical reduction

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Felix J Berger et al.
ACS nano, 13(8), 9259-9269 (2019-08-06)
The functionalization of semiconducting single-walled carbon nanotubes (SWNTs) with sp3 defects that act as luminescent exciton traps is a powerful means to enhance their photoluminescence quantum yield (PLQY) and to add optical properties. However, the synthetic methods employed to introduce
Denham, M. and Ingham, R.K.
The Journal of Organic Chemistry, 23, 1298-1298 (1958)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service