Skip to Content
Merck
All Photos(4)

Documents

A8509

Sigma-Aldrich

Lithium acetoacetate

≥90% (HPLC)

Synonym(s):

Acetoacetic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOLi
CAS Number:
Molecular Weight:
108.02
Beilstein:
5774841
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Li+].CC(=O)CC([O-])=O

InChI

1S/C4H6O3.Li/c1-3(5)2-4(6)7;/h2H2,1H3,(H,6,7);/q;+1/p-1

InChI key

UTLRZTUJSMCBHB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium acetoacetate can be used as an acetoacetate standard for the measurement of acetoacetate in biological samples. It can also be used for the preparation of sodium acetoacetate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cristian Palmiere et al.
Legal medicine (Tokyo, Japan), 14(1), 17-20 (2011-12-20)
Isopropyl alcohol (IPA) is widely used as an industrial solvent and cleaning fluid. After ingestion or absorption, IPA is converted into acetone by alcohol dehydrogenase. However, in ketosis, acetone can be reduced to IPA. The aim of this study was
Preparation of crystalline lithium acetoacetate.
Hall LM
Analytical Biochemistry, 3(1), 75-80 (1962)
Bei Han et al.
Applied microbiology and biotechnology, 91(3), 565-576 (2011-05-03)
Development of a butanologenic strain with high selectivity for butanol production is often proposed as a possible route for improving the economics of biobutanol production by solventogenic Clostridium species. The acetoacetate decarboxylase (aadc) gene encoding acetoacetate decarboxylase (AADC), which catalyzes
Eun Ae Jeong et al.
Experimental neurology, 232(2), 195-202 (2011-09-24)
Similar to fasting, the ketogenic diet (KD) has anti-inflammatory effects and protects against excitotoxicity-mediated neuronal cell death. Recent studies have shown that peroxisome proliferator-activated receptor (PPAR)γ has anti-inflammatory effects in seizure animal models. However, the exact mechanisms underlying the anti-inflammatory
Hao Liang et al.
Journal of Cancer, 10(16), 3735-3745 (2019-07-25)
BDH2 is a short-chain dehydrogenase/reductase family member involved in several biological and pathological processes, including the utilization of cytosolic ketone bodies, immunocyte regulation and tumor progression. In this study, we first revealed that BDH2 was downregulated in HCC tissues by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service