Skip to Content
Merck
All Photos(1)

Key Documents

D31206

Sigma-Aldrich

Dibenz[a,c]anthracene

97%

Synonym(s):

1,2:3,4-Dibenzanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
1912415
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
97%

Quality Level

Assay

97%

form

powder

bp

518 °C (lit.)

mp

205-207 °C (lit.)

SMILES string

c1ccc2cc3c4ccccc4c5ccccc5c3cc2c1

InChI

1S/C22H14/c1-2-8-16-14-22-20-12-6-4-10-18(20)17-9-3-5-11-19(17)21(22)13-15(16)7-1/h1-14H

InChI key

RAASUWZPTOJQAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Chouroulinkov et al.
Cancer letters, 19(1), 21-26 (1983-05-01)
The tumor-initiating activities of dibenz[a,c]anthracene (DBA) and of the related trans-1,2-, 3,4- and 10,11-dihydrodiols have been examined on mouse skin subsequently promoted with 12-O-tetradecanoyl-phorbol-13-acetate (TPA). The 1,2- and 10,11-dihydrodiols were active and were more active than equivalent doses of either
M Hall et al.
Carcinogenesis, 9(5), 865-868 (1988-05-01)
Incubation of r-1,t-2-dihydroxy-t-3,4-oxy-1,2,3,4-tetrahydrochrysene (anti-chrysene-1,2-diol 3,4-oxide), the bay-region diol-epoxide of chrysene, with rat liver microsomes in the presence of NADP+ and DNA, followed by 32P-postlabelling analysis of the DNA, revealed the presence of at least two adducts not detected when anti-chrysene-1,2-diol
Dibenz[a,c]anthracene.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 32, 289-297 (1983-12-01)
S Kumar et al.
Mutation research, 242(4), 337-343 (1990-12-01)
The mutagenic activities of dibenz[a,c]anthracene (DB[a,c]A), and its 11 derivatives, including 3 diols, 6 phenols and 2 oxepines, were studied in the TA100 strain of Salmonella typhimurium at doses varying from 0 to 20 micrograms/plate in the presence of a
A Hewer et al.
Carcinogenesis, 2(12), 1345-1352 (1981-01-01)
In rat liver microsomal preparations, the 10,11-dihydrodiol of dibenz[a,c]anthracene (DBA) is metabolized to r-10,t-11-dihydroxy-t-12,13-oxy-10,11,12,13-tetrahydrodibenz[a,c]anthracene (anti-DBA 10,11-diol 12,13-oxide), the anti isomer of a non-bay-region diol-epoxide of DBA. When 3H-labelled DBA or trans-10,11-dihydro-10,11-dihydroxydibenz[a,c]anthracene were metabolized in this system in the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service