Skip to Content
Merck
All Photos(2)

Key Documents

91861

Supelco

Dibenz[a,h]anthracene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1,2:5,6-Dibenzanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
1912416
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

524 °C (lit.)

mp

262-265 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

c1ccc2c(c1)ccc3cc4c(ccc5ccccc45)cc23

InChI

1S/C22H14/c1-3-7-19-15(5-1)9-11-17-14-22-18(13-21(17)19)12-10-16-6-2-4-8-20(16)22/h1-14H

InChI key

LHRCREOYAASXPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ion Mobility: TWCCSN2 value of 158.3 Å2 [M+H]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 91861 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A L Juhasz et al.
Letters in applied microbiology, 30(5), 396-401 (2000-05-03)
The ability of Stenotrophomonas maltophilia strain VUN 10,003 to degrade and detoxify high molecular weight polycyclic aromatic hydrocarbons (PAHs) was evaluated in a basal liquid medium. Using high cell density inocula of strain VUN 10,003, the concentration of pyrene, fluoranthene
B Z Zhong et al.
Mutation research, 326(2), 147-153 (1995-02-01)
Benz[a]anthracene (BA), dibenz[a,h]anthracene (DBA) and dibenzo[a,i]pyrene (DBP) are polycyclic aromatic hydrocarbons (PAHs) found in incomplete combustion products of fossil fuels, coal tar, and other organic materials. Workers in related industries may be exposed to these chemicals by inhalation. The information
M Shou et al.
Archives of biochemistry and biophysics, 328(1), 201-207 (1996-04-01)
The metabolic activation of the potent carcinogen dibenzo[a,h]anthracene (DB[a,h]A) was investigated with recombinant human cytochrome P450 enzymes 1A2, 2B6, 2C8, 2C9, 2E1, 3A3, 3A4, and 3A5 expressed in hepatoma G2 cells and with 14 different human liver microsomes. Three dihydrodiols
A Perico et al.
Archives of environmental health, 56(6), 506-512 (2002-04-18)
In this study, the authors evaluated exposure to airborne polycyclic aromatic hydrocarbons (PAHs) in workers exposed to exhaust gas from cars, and they assessed the efficiency of urinary 1-hydroxypyrene as an indicator of exposure to pyrene and PAHs. The authors
Heinz Frank et al.
Chemical research in toxicology, 24(12), 2258-2268 (2011-11-18)
The synthesis of the 8 possible stereoisomeric diol epoxides (DEs) at the terminal benzo ring of carcinogenic dibenz[a,h]anthracene (DBA) is reported. trans-3,4-Dihydroxy-3,4-dihydro-DBA (1) afforded the 4 bay region DEs: the enantiomeric pairs of the anti diastereomers (+)-3/(-)-3 and of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service