Skip to Content
Merck
All Photos(1)

Documents

D222003

Sigma-Aldrich

Dodecyl aldehyde

92%

Synonym(s):

Lauric aldehyde, Aldehyde C12, Dodecanal, Lauraldehyde, Laurinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CHO
CAS Number:
Molecular Weight:
184.32
Beilstein:
1703917
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

92%

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

185 °C/100 mmHg (lit.)

density

0.831 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC=O

InChI

1S/C12H24O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h12H,2-11H2,1H3

InChI key

HFJRKMMYBMWEAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Gianna Panasia et al.
Applied and environmental microbiology, 84(13) (2018-04-22)
The opportunistic pathogen Pseudomonas aeruginosa strain PAO1 is able to use a variety of organic pollutants as growth substrates, including the anionic detergent sodium dodecyl sulfate (SDS) and long-chain alkanes. While the enzymes initiating SDS and alkane degradation are well
Markus A Keller et al.
Nature communications, 5, 4439-4439 (2014-07-23)
Mutations in the gene coding for membrane-bound fatty aldehyde dehydrogenase (FALDH) lead to toxic accumulation of lipid species and development of the Sjögren-Larsson Syndrome (SLS), a rare disorder characterized by skin defects and mental retardation. Here, we present the crystallographic
James Liu et al.
Journal of pharmaceutical sciences, 104(3), 1187-1196 (2015-01-13)
The objective of this study is to develop and compare several Sorafenib-loaded biocompatible nanoparticle models in order to optimize drug delivery and tumor cellular kill thereby improving the quality of Sorafenib-regimented chemotherapy. Sorafenib-loaded poly (lactic-co-glycolic) acid (PLGA), 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) liposomes
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service