Skip to Content
Merck
All Photos(1)

Key Documents

D221600

Sigma-Aldrich

1-Dodecene

95%

Synonym(s):

n-Dodec-1-ene, alpha-Dodecene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH=CH2
CAS Number:
Molecular Weight:
168.32
Beilstein:
1699848
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.8 (vs air)

Quality Level

vapor pressure

0.2 mmHg ( 20 °C)

Assay

95%

form

liquid

autoignition temp.

491 °F

expl. lim.

~4.7 %

impurities

≤2.0% 2-butyl-1-octene
≤2.0% 2-ethyl-1-decene
≤2.0% n-dodecane

refractive index

n20/D 1.429 (lit.)

bp

214-216 °C (lit.)

mp

−35 °C (lit.)

density

0.758 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCC=C

InChI

1S/C12H24/c1-3-5-7-9-11-12-10-8-6-4-2/h3H,1,4-12H2,2H3

InChI key

CRSBERNSMYQZNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Dodecene can be used as a substrate in:
  • Preparation of linear phenyldodecanes by reacting with benzene using various acid catalysts.
  • Hydroformylation reaction by microemulsion method in the presence of rhodium catalyst.
  • Synthesis of synthetic lubricants using chromium catalysts via trimerization.
  • Alkylation of toluene
  • Polymerization using various supported catalysts.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroformylation of 1-dodecene using Rh-TPPTS in a microemulsion
Haumann M, et al.
Applied Catalysis A: General, 225(1-2), 239-249 (2002)
Polymerization of ethene and longer chained olefins by metallocene catalysis
Macromolecular Symposia, 226(1), 25-34 (2005)
Synthesis of Synthetic Lubricants by Trimerization of 1-Decene and 1-Dodecene with Homogeneous Chromium Catalysts
Wasserscheid P, et al.
Advanced Synthesis & Catalysis, 343(8), 814-818 (2001)
Synthesis of linear phenyldodecanes by the alkylation of benzene with 1-dodecene over non-zeolitic catalysts
Yadav GD and Doshi NS
Organic Process Research & Development, 6(3), 263-272 (2002)
Synthesis and characterization of HPW/MCM-41 (Si) and HPW/MCM-41 (Si/Al) catalysts: Activity for toluene alkylation with 1-dodecene
Llanos A, et al.
Catalysis Today, 133(8), 20-27 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service