Recommended Products
vapor density
4.84 (vs air)
Quality Level
vapor pressure
1.6 mmHg ( 23.8 °C)
Assay
≥97.0% (GC)
form
liquid
autoignition temp.
455 °F
refractive index
n20/D 1.421 (lit.)
bp
166.5-173.5 °C (lit.)
mp
−66.3-−66 °C (lit.)
density
0.741 g/mL at 25 °C (lit.)
functional group
allyl
SMILES string
CCCCCCCCC=C
InChI
1S/C10H20/c1-3-5-7-9-10-8-6-4-2/h3H,1,4-10H2,2H3
InChI key
AFFLGGQVNFXPEV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
1-Decene is a marker of oxidative stress. The reactivity ratio for the copolymerization of 1-decene with 2/MAO (methylaluminoxane) was studied.
Application
1-Decene was used to study the hydrosilylation kinetics of hydride-capped silicon nanocrystals.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
111.2 °F - closed cup
Flash Point(C)
44.0 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Dalton transactions (Cambridge, England : 2003), 42(25), 9139-9147 (2013-03-15)
Several analogues of the sterically expanded constrained geometry catalyst Me2Si(η(1)-C29H36)(η(1)-N-tBu)ZrCl2·OEt2 (2) were synthesized to assess the effect on branching and molecular weight for ethylene homopolymerization. Catalysts based on tetramethyltetrahydrobenzofluorene (TetH), ethylTetH, t-butylTetH, and octamethyloctahydrodibenzofluorenyl (OctH) bearing a diphenylsilyl bridge were
Chemical communications (Cambridge, England), 49(97), 11361-11363 (2013-10-29)
Hydrosilylation kinetics of hydride-capped silicon nanocrystals with 1-decene are reported for the first time. In a side-by-side comparison, it is found that microwave heating has no evident acceleration effect on the hydrosilylation rate relative to conventional thermal heating.
A122: metabolomic analysis of breath volatile organic compounds reveals unique breathprints in children with juvenile idiopathic arthritis.
Arthritis & Rheumatology (Hoboken, N.J.), 66 Suppl 11, S159-S159 (2014)
PLoS computational biology, 11(10), e1004368-e1004368 (2015-10-28)
The knowledge of multiple conformational states is a prerequisite to understand the function of membrane transport proteins. Unfortunately, the determination of detailed atomic structures for all these functionally important conformational states with conventional high-resolution approaches is often difficult and unsuccessful.
Proceedings of the National Academy of Sciences of the United States of America, 111(51), 18381-18386 (2014-12-10)
Excitation-evoked Ca(2+) influx is the fastest and most ubiquitous chemical trigger for cellular processes, including neurotransmitter release, muscle contraction, and gene expression. The voltage dependence and timing of Ca(2+) entry are thought to be functions of voltage-gated calcium (CaV) channels
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service