Skip to Content
Merck
All Photos(1)

Key Documents

D209317

Sigma-Aldrich

Diphenylmethane

99%

Synonym(s):

Benzylbenzene, Methylenedibenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CH2
CAS Number:
Molecular Weight:
168.23
Beilstein:
1904982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.79 (vs air)

vapor pressure

<1 mmHg ( 77 °C)

Assay

99%

form

liquid

autoignition temp.

905 °F

refractive index

n20/D 1.577 (lit.)

bp

264 °C (lit.)

mp

22-24 °C (lit.)

density

1.006 g/mL at 25 °C (lit.)

SMILES string

C(c1ccccc1)c2ccccc2

InChI

1S/C13H12/c1-3-7-12(8-4-1)11-13-9-5-2-6-10-13/h1-10H,11H2

InChI key

CZZYITDELCSZES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diphenylmethane is an aromatic compound used as a key building block in the synthesis of elastase inhibitors.

Application

  • Diphenylmethane is widely used in the synthesis of luminogens for aggregation-induced emission (AIE).
  • It is used in the preparation of a polymerization initiator, diphenylmethyl potassium (DPMK).
  • It is one of the precursors in the synthesis of a dendrimeric polycyclic aromatic hydrocarbon (PAH), hexakis[4-(1,1,2-triphenyl-ethenyl)phenyl]benzene.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

266.0 °F

Flash Point(C)

130 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of functionalized aromatic oligomers from a versatile diphenylmethane template
JG Bruno et al.
The Journal of Organic Chemistry, 62, 5174-5190 (1997)
Real-time monitoring of cell apoptosis and drug screening using fluorescent light-up probe with aggregation-induced emission characteristics.
Shi H, et al.
Journal of the American Chemical Society, 134(43), 17972-17981 (2012)
Hopping of a single hole in hexakis [4-(1, 1, 2-triphenyl-ethenyl) phenyl] benzene cation radical through the hexaphenylbenzene propeller.
Rathore R, et al.
Organic Letters, 6(11), 1689-1692 (2004)
The theoretical conformational analysis of some bridged aromatic compounds: diphenyl ether, diphenylmethane, benzophenone and diphenyl sulphide
VA Zubkov et al.
Journal of Molecular Structure, 27, 139-149 (1975)
Synthesis and surface properties of amphiphilic star-shaped and dendrimer-like copolymers based on polystyrene core and poly (ethylene oxide) corona.
Francis R, et al.
Macromolecules, 36(22), 8253-8259 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service