Skip to Content
Merck
All Photos(3)

Documents

D208000

Sigma-Aldrich

1,6-Diphenyl-1,3,5-hexatriene

98%

Synonym(s):

DPH, Dicinnamyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH=CH)3C6H5
CAS Number:
Molecular Weight:
232.32
Beilstein:
1907440
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

mp

199-203 °C (lit.)

fluorescence

λex 350 nm; λem 452 nm in methanol

SMILES string

C(=C/C=C/c1ccccc1)\C=C\c2ccccc2

InChI

1S/C18H16/c1(5-11-17-13-7-3-8-14-17)2-6-12-18-15-9-4-10-16-18/h1-16H/b2-1+,11-5+,12-6+

InChI key

BOBLSBAZCVBABY-WPWUJOAOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DPH is a lipophilic molecule used to study membrane fluidity and polarization by measuring its fluorescence properties.

Application

Used an alternative to Nile red for fluorescent detection of lipid droplets

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temperature dependence of 1, 6-diphenyl-1, 3, 5-hexatriene fluorescence in phospholipid artificial membranes
Andrich MP, et al.
Biochemistry, 15, 1257-1261 (1976)
Florie Desriac et al.
Pathogens (Basel, Switzerland), 7(2) (2018-04-27)
We have previously shown that the C-type Natriuretic Peptide (CNP), a peptide produced by lungs, is able to impact Pseudomonasaeruginosa physiology. In the present work, the effect of CNP at different concentrations on P. aeruginosa biofilm formation was studied and
Markku Hakala et al.
The Journal of biological chemistry, 293(13), 4818-4829 (2018-02-10)
Membrane phosphoinositides control organization and dynamics of the actin cytoskeleton by regulating the activities of several key actin-binding proteins. Twinfilin is an evolutionarily conserved protein that contributes to cytoskeletal dynamics by interacting with actin monomers, filaments, and the heterodimeric capping
Yaser Kashcooli et al.
Biomacromolecules, 17(11), 3838-3844 (2016-10-12)
Layer-by-layer deposition of polyelectrolytes (PEs) onto self-assembled liposomes represents an alternative to PE deposition on solid particles for the formation of hollow nanoscale capsules. This work examines how competition between PE-liposome and inter-PE interactions drives the structure and colloidal stability
Natalie Spang et al.
Autophagy, 10(12), 2297-2309 (2014-12-17)
Macroautophagy is a degradative pathway that sequesters and transports cytosolic cargo in autophagosomes to lysosomes, and its deterioration affects intracellular proteostasis. Membrane dynamics accompanying autophagy are mostly elusive and depend on trafficking processes. RAB GTPase activating proteins (RABGAPs) are important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service