D187208
2,5-Dimethyltetrahydrofuran, mixture of cis and trans
96%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12O
CAS Number:
Molecular Weight:
100.16
Beilstein:
102563
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
96%
form
liquid
refractive index
n20/D 1.404 (lit.)
bp
90-92 °C (lit.)
density
0.833 g/mL at 25 °C (lit.)
SMILES string
CC1CCC(C)O1
InChI
1S/C6H12O/c1-5-3-4-6(2)7-5/h5-6H,3-4H2,1-2H3
InChI key
OXMIDRBAFOEOQT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
80.6 °F - closed cup
Flash Point(C)
27 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A S Anderson et al.
The Journal of organic chemistry, 65(15), 4648-4654 (2000-08-26)
2'-Deoxy-5-methyleneuridin-5-yl (1) is produced in a variety of DNA damage processes and is believed to result in the formation of lesions that are mutagenic and refractory to enzymatic repair. 2'-Deoxy-5-methyleneuridin-5-yl (1) was independently generated under anaerobic conditions via Norrish Type
John M Simmie
The journal of physical chemistry. A, 116(18), 4528-4538 (2012-04-13)
The enthalpies of formation, entropies, specific heats at constant pressure, enthalpy functions, and all carbon-hydrogen and carbon-methyl bond dissociation energies have been computed using high-level methods for the cyclic ethers (oxolanes) tetrahydrofuran, 2-methyltetrahydrofuran, and 2,5-dimethyltetrahydrofuran. Barrier heights for hydrogen-abstraction reactions
Weiran Yang et al.
ChemSusChem, 3(5), 597-603 (2010-05-04)
Existing technologies to produce liquid fuels from biomass are typically energy-intensive, multistep processes. Many of these processes use edible biomass as starting material. Carbohydrates, such as mono- and polysaccharides and cellulose, typically constitute 50-80% of plant biomass. Herein, we report
M S Akhlaq et al.
International journal of radiation biology and related studies in physics, chemistry, and medicine, 51(1), 91-102 (1987-01-01)
Thiyl radicals (RS) formed by the reaction of radiolytically generated OH radicals with thiols, e.g. 1,4-dithiothreitol (DTT), react with cis- and trans-2,5-dimethyltetrahydrofuran by abstracting an H atom in the alpha-position to the ether function (k approximately equal to 5 X
Matthew R Grochowski et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(39), 12363-12371 (2012-08-24)
Carbohydrates, such as fructose, can be fully dehydroxylated to 2,5-dimethyltetrahydrofuran (DMTHF), a valuable chemical and potential gasoline substitute, by the use of a dual catalytic system consisting of HI and RhX(3) (X=Cl, I). A mechanistic study has been carried out
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service