Skip to Content
Merck
All Photos(2)

Documents

137316

Sigma-Aldrich

5-Methylfurfural

ReagentPlus®, 99%

Synonym(s):

5-Methyl-2-furaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
Beilstein:
106895
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.531 (lit.)

bp

187 °C (lit.)

density

1.107 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccc(C)o1

InChI

1S/C6H6O2/c1-5-2-3-6(4-7)8-5/h2-4H,1H3

InChI key

OUDFNZMQXZILJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methylfurfural is formed during the photoexposition of ranitidine hydrochloride. It is employed as potential age marker for Madeira wine. It is a volatile compound present in Lavandula stoechas, Lavandula angustifolia and Lavandula angustifolia x latifolia unifloral honeys.

Application

5-Methylfurfural was used to investigate the volatile compounds isolated from Erica arborea and Calluna vulgaris unifloral honeys.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Floral quality and discrimination of Lavandula stoechas, Lavandula angustifolia and Lavandula angustifolia x latifolia honeys.
Guyot-Declerck C, et al.
Food Chemistry, 79(4), 453-459 (2002)
Rosa Perestrelo et al.
Journal of agricultural and food chemistry, 59(7), 3186-3204 (2011-03-08)
The establishment of potential age markers of Madeira wine is of paramount significance as it may contribute to detect frauds and to ensure the authenticity of wine. Considering the chemical groups of furans, lactones, volatile phenols, and acetals, 103 volatile
E M Jouad et al.
Journal of inorganic biochemistry, 86(2-3), 565-571 (2001-09-22)
5-Methyl 2-furfuraldehyde thiosemicarbazone (M5HFTSC) with nickel(II) leads to three types of complexes: [Ni(M5HFTSC)(2)X(2)], [Ni(M5FTSC)(2)] and [Ni(M5FTSC)(2)] x 2DMF. In the first type the ligand remains in thione form, while in the two other, the anionic thiolato form is involved. The
Floral origin markers of heather honeys: Erica arborea and Calluna vulgaris.
Guyot C, et al.
Food Chemistry, 64(1), 3-11 (1999)
S Uddin et al.
Biochemistry and molecular biology international, 35(1), 185-195 (1995-01-01)
Furfural and methylfurfural are dietary mutagens and are present in various food products and beverages. The alkaline-induced unwinding of calf thymus DNA permitted the measurement of the number of strand breaks formed by furfural and methylfurfural, as a function of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service