D141003
2-(Dimethylamino)ethanethiol hydrochloride
95%
Synonym(s):
MEDA
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
(CH3)2NCH2CH2SH · HCl
CAS Number:
Molecular Weight:
141.66
Beilstein:
3906670
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
solid
mp
150-160 °C (lit.)
SMILES string
Cl.CN(C)CCS
InChI
1S/C4H11NS.ClH/c1-5(2)3-4-6;/h6H,3-4H2,1-2H3;1H
InChI key
NRVFDGZJTPCULU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
2-(Dimethylamino)ethanethiol hydrochloride can be used:
- To synthesize fluorescent thiocholine esters containing dansyl group in the acyl portions, which are potent substrates for the human fibrinoligase.
- As a cationic ligand for the surface modification of gold nanoparticles.
- For the modification of cellulose 10-undecenoyl ester (CUE) to form organo-soluble ionic cellulose derivatives.
Metal-complexing agent
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Peyman Hadji et al.
Journal of bone oncology, 7, 1-12 (2017-04-18)
Several guidelines have been reported for bone-directed treatment in women with early breast cancer (EBC) for averting fractures, particularly during aromatase inhibitor (AI) therapy. Recently, a number of studies on additional fracture related risk factors, new treatment options as well
Methods report: European S3-Guideline on the systemic treatment of psoriasis vulgaris - Update Apremilast and Secukinumab - EDF in cooperation with EADV and IPC.
C Dressler et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 31(12), 1964-1977 (2017-09-26)
Small, water-soluble, ligand-stabilized gold nanoparticles synthesized by interfacial ligand exchange reactions
Warner MG
Chemistry of Materials, 12(11), 3316-3320 (2000)
Fabian Uddén et al.
mSphere, 5(1) (2020-02-07)
Naturally acquired opsonic antipneumococcal antibodies are commonly found in nonvaccinated adults and confer protection against infection and colonization. Despite this, only limited data exist regarding the adaptive immune response after pneumococcal exposure. To investigate the dynamics of naturally acquired antipneumococcal
Stimuli-responsive nanoparticles from ionic cellulose derivatives
Wang Y
Nanoscale, 8(1), 648-657 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service