Skip to Content
Merck
All Photos(1)

Key Documents

304255

Sigma-Aldrich

3-Chloro-3-methylpentane

97%

Synonym(s):

1-Ethyl-1-methylpropyl chloride, 3-Chloro-3-methylpentane, 3-Methyl-3-chloropentane, Diethylmethylcarbinyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(C2H5)2Cl
CAS Number:
Molecular Weight:
120.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

115-116 °C/760 mmHg (lit.)

density

0.885 g/mL at 25 °C (lit.)

functional group

alkyl halide
chloro

SMILES string

CCC(C)(Cl)CC

InChI

1S/C6H13Cl/c1-4-6(3,7)5-2/h4-5H2,1-3H3

InChI key

SGWJUIFOPCZXMR-UHFFFAOYSA-N

Related Categories

Application

3-Chloro-3-methylpentane(3C3MP) has structure similar to tertiary chloride groups which are formed as structural defects during the radical polymerization of vinyl chloride (VC). Therefore, 3C3MP is an ideal model compound to study if tertiary chloride defect present in commercial PVC can be active initiating species for metal-catalyzed living radical graft copolymerization.
3C3MP can also be used as a reactant to prepare chloro(1-ethyl-1-methylpropyl)magnesium (Grignard reagent), which is used in the Kumada–Tamao–Corriu coupling reaction.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An N-heterocyclic carbene-based nickel catalyst for the Kumada-Tamao-Corriu coupling of aryl bromides and tertiary alkyl Grignard reagents
Ando S, et al.
Tetrahedron Letters, 57(30), 3287-3290 (2016)
The enthalpies of solution of 2-chloro-2-methylpropane, 2-chloro-2-methylbutane and 3-chloro-3-methylpentane in mono-and dialcohols at 298.15 K.
Albuquerque LMPC, et al.
Thermochimica Acta, 275(1), 67-73 (1996)
Metal-catalyzed living radical graft copolymerization of olefins initiated from the structural defects of poly (vinyl chloride)
Percec V and Asgarzadeh F
Journal of Polymer Science Part A: Polymer Chemistry, 39(7), 1120-1135 (2001)
Kinetic and thermodynamic study of 2-bromo-2-methylbutane, 2-chloro-2-methylbutane and 3-chloro-3-methylpentane in diols.
Albuquerque LC, et al.
Journal of Physical Organic Chemistry, 11(1), 36-40 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service