Recommended Products
form
solid
SMILES string
[Na+].OCC([O-])=O
InChI
1S/C2H4O3.Na/c3-1-2(4)5;/h3H,1H2,(H,4,5);/q;+1/p-1
InChI key
VILMUCRZVVVJCA-UHFFFAOYSA-M
Related Categories
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pharmaceutical development and technology, 21(2), 222-230 (2014-12-19)
The effect of lactose particle size on the extent of pellet coat damage was investigated. The extent of pellet coat damage increased linearly with lactose median particle size. It was observed that coated pellets compressed with coarser lactose grades had
Journal of young pharmacists : JYP, 3(3), 211-215 (2011-09-08)
In the present study, fast disintegrating tablets were designed with a view to enhance patient compliance. In this method, the hull of Hordeum vulgare, cross carmellose sodium, and sodium starch glycolate were used as superdisintegrants (4 and 6%), along with
Chemical & pharmaceutical bulletin, 63(4), 295-299 (2015-04-04)
In order to enhance the dissolution rate of dronedarone hydrochloride (DRN), a novel Soluplus(®) (polyethyleneglycol-polyvinyl caprolactam-polyvinyl acetate grafted copolymer)-based solid dispersion (SD) was formulated using a hot melt extrusion technique. The physical characteristics determined using scanning electron microscopy and X-ray
Indian journal of pharmaceutical sciences, 70(2), 180-185 (2008-03-01)
Aceclofenac, a non-steroidal antiinflammatory drug, is used for posttraumatic pain and rheumatoid arthritis. Aceclofenac fast-dispersible tablets have been prepared by direct compression method. Effect of superdisintegrants (such as, croscarmellose sodium, sodium starch glycolate and crospovidone) on wetting time, disintegration time
Indian journal of pharmaceutical sciences, 70(2), 203-207 (2008-03-01)
The present study is aimed at the overall improvement in the efficacy, reduced toxicity and enhancement of therapeutic index of cisplatin. Solid lipid nanoparticulate delivery system of cisplatin has been developed by microemulsification method by using stearic acid, soy lecithin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service