Skip to Content
Merck
All Photos(4)

Documents

T0632

Sigma-Aldrich

Sodium thioglycolate

≥96.5% (iodometric)

Synonym(s):

Mercaptoacetic acid sodium salt, Thioglycolic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COONa
CAS Number:
Molecular Weight:
114.10
Beilstein:
4569109
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥96.5% (iodometric)

form

powder

color

white to off-white

useful pH range

6.7 (20 °C, 100 g/L)

mp

>300 °C (lit.)

solubility

water: ~609.1 g/L at 20 °C (OECD Test Guideline 105)

storage temp.

−20°C

SMILES string

[Na+].[O-]C(=O)CS

InChI

1S/C2H4O2S.Na/c3-2(4)1-5;/h5H,1H2,(H,3,4);/q;+1/p-1

InChI key

GNBVPFITFYNRCN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium thioglycolate has been used:
  • as a growth supplement in enrichment media and to study its effect on Arcobacter
  • in influenza hemagglutinin formulation in order to reduce disulfide-mediated cross-linking and early potency loss
  • in electron microscopy

Biochem/physiol Actions

Sodium thioglycolate acts as reducing agent and is suitable for anaerobic and microaerophilic bacterial growth.
Sodium thioglycolate is a commonly used reagent for bacteriological research to maintain reducing conditions in media. Thioglycolate can also protect enzymes against inactivation by maintaining protein thiol groups in the reduced state. Thioglycolate medium is frequently used in inflammation research to elicit a neutrophil and macrophage response in vivo.

Caution

At room temperature, concentrations over 70% in water tend to form 1-2% thioglycolides per month which hydrolyze to the original free compound when made acidic or alkaline. The 70% solution oxidizes in air but is stable at room temperature when tightly closed. Thioglycolate salts may also lose purity on storage. The exclusion of air does not materially improve stability.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Met. Corr. 1 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuya Omotezako et al.
Scientific reports, 7, 44226-44226 (2017-03-11)
The maternal contribution to the oocyte cytoplasm plays an important role during embryogenesis because it is involved in early cell fate specification and embryonic axis establishment. However, screening projects targeting maternal factors have only been conducted in a limited number
Development of Parvalbumin-Expressing Basket Terminals in Layer II of the Rat Medial Entorhinal Cortex
Berggaard N, et al.
eNeuro, 5(3) (2018)
Clinical Ophthalmology: Contemporary Perspectives - E-Book, 176-176 (2012)
Kim De Paepe et al.
PeerJ, 7, e6293-e6293 (2019-02-01)
Undigestible, insoluble food particles, such as wheat bran, are important dietary constituents that serve as a fermentation substrate for the human gut microbiota. The first step in wheat bran fermentation involves the poorly studied solubilization of fibers from the complex
Kouhei Oonuma et al.
Developmental biology, 420(1), 178-185 (2016-10-30)
The Ciona intestinalis larva has two distinct photoreceptor organs, a conventional pigmented ocellus and a nonpigmented ocellus, that are asymmetrically situated in the brain. The ciliary photoreceptor cells of these ocelli resemble visual cells of the vertebrate retina. Precise elucidation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service