BR4640240
BRAND® Dispensette® S Trace Analysis Analog-adjustable bottle-top dispenser
tantalum spring, volume range 1-10 mL, without recirculation valve
Synonym(s):
bottle top dispensers
All Photos(1)
About This Item
Recommended Products
feature
incremental 0.2 mL
without recirculation valve
manufacturer/tradename
BRAND 4640240
volume range
1-10 mL
fitting
45 mm
General description
Dispensette S Trace Analysis is specialized for dispensing high-purity acids, bases and H2O2.
Also suitable for dispensing of hydrofluoric acid when using platinum-iridium valve spring.
Also suitable for dispensing of hydrofluoric acid when using platinum-iridium valve spring.
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Other Notes
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)