Key Documents
BR4630150
BRAND® Dispensette® S Organic, Analog-adjustable bottle-top dispenser
volume range 2.5-25 mL, without recirculation valve
Synonym(s):
bottle top dispensers
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
incremental 0.5 mL
DE-M marking
without recirculation valve
manufacturer/tradename
BRAND 4630150
volume range
2.5-25 mL
fitting
(GL 32-33)
GL 38
S 40
General description
Dispensette S Organic is ideal for dispensing of organic solvents and acids
Features and Benefits
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Other Notes
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)