Skip to Content
Merck
All Photos(2)

Key Documents

B99006

Sigma-Aldrich

2-sec-Butylphenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)C6H4OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1210026
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.03 mmHg ( 20 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.522 (lit.)

bp

226-228 °C (lit.)

mp

12 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

SMILES string

CCC(C)c1ccccc1O

InChI

1S/C10H14O/c1-3-8(2)9-6-4-5-7-10(9)11/h4-8,11H,3H2,1-2H3

InChI key

NGFPWHGISWUQOI-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

2-sec-Butylphenol is a precursor to synthesize various coumarin and benzofuran derivatives that are used as lipid-lowering agents and potential bone anabolic agents. It can also be used as a solvent in organic synthesis.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzofuran-pyran hybrids: A new class of potential bone anabolic agents.
Gupta S, et al.
Bioorganic & Medicinal Chemistry Letters, 28(10), 1719-1724 (2018)
Benzofuran-dihydropyridine hybrids: A new class of potential bone anabolic agents.
Modukuri R K, et al.
Bioorganic & Medicinal Chemistry, 25(24), 6450-6466 (2017)
Discovery of coumarin?dihydropyridine hybrids as bone anabolic agents.
Sashidhara K V, et al.
Journal of Medicinal Chemistry, 56(1), 109-122 (2012)
Spectrophotometric determination of nitrate in vegetable products using 2-sec-butylphenol.
A Tanaka et al.
The Analyst, 107(1271), 190-194 (1982-02-01)
N E Hopkins et al.
Biochemical and biophysical research communications, 244(3), 868-872 (1998-04-16)
CYP102 (Cytochrome P450BM-3) is induced in Bacillus megaterium by barbiturates, peroxisome proliferators, and nonsteroidal anti-inflammatory drugs. We now describe the induction of CYP102 in B. megaterium by 17 beta-estradiol and by 4-sec-butylphenol. These estrogens interact with the repressor protein Bm3R1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service