Skip to Content
Merck
All Photos(1)

Key Documents

B9750

Sigma-Aldrich

Benzophenone-3,3′,4,4′-tetracarboxylic dianhydride

98%

Synonym(s):

4,4′-Carbonyldiphthalic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H6O7
CAS Number:
Molecular Weight:
322.23
Beilstein:
761777
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor density

1.4 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 0 °C)

Assay

98%

form

powder

autoignition temp.

975 °F

expl. lim.

16 %

mp

218-222 °C (lit.)

SMILES string

O=C1OC(=O)c2cc(ccc12)C(=O)c3ccc4C(=O)OC(=O)c4c3

InChI

1S/C17H6O7/c18-13(7-1-3-9-11(5-7)16(21)23-14(9)19)8-2-4-10-12(6-8)17(22)24-15(10)20/h1-6H

InChI key

VQVIHDPBMFABCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzophenone-3,3′,4,4′-tetracarboxylic acid is formed by hydrolysis, it can be converted to the dianhydride by treating with Ac2O (molar ratio 4 to 1 of acid).[1]The dianhydride is used to synthesize polyimides, which are highly flexible in nature due to the presence of keto and carbonyl groups in the dianhydride molecule. The keto and carbonyl groups subsequently increase the spacing between the imide rings enhancing the solubility and processibility.

Application

Used in the synthesis of polyimides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Armarego WLF and Chai C
Purification of Laboratory Chemicals (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service