Skip to Content
Merck
All Photos(1)

Documents

B8002

Sigma-Aldrich

2,3-Benzofuran

99%

Synonym(s):

Coumarone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O
CAS Number:
Molecular Weight:
118.13
Beilstein:
107704
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.566 (lit.)

bp

173-175 °C (lit.)

density

1.072 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

c1ccc2occc2c1

InChI

1S/C8H6O/c1-2-4-8-7(3-1)5-6-9-8/h1-6H

InChI key

IANQTJSKSUMEQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miyuki Yamaguchi et al.
The Journal of organic chemistry, 78(18), 9270-9281 (2013-08-21)
A dihydroxyterphenylphosphine bearing cyclohexyl groups on the phosphorus atom (Cy-DHTP) was found to be a powerful ligand for the palladium-catalyzed one-pot synthesis of substituted benzo[b]furans from 2-chlorophenols and terminal alkynes. This catalyst system was also applicable to the sequential one-pot
Stefano Protti et al.
The Journal of organic chemistry, 77(15), 6473-6479 (2012-07-11)
2-Substituted benzo[b]furans were synthesized by a one-step metal-free photochemical reaction between 2-chlorophenol derivatives and terminal alkynes by tandem formation of an aryl-C and a C-O bond via an aryl cation intermediate. The mild conditions and the application to chlorophenols rather
Ulrika Olofsson et al.
Chemosphere, 90(1), 28-35 (2012-08-28)
A screening of metals, persistent organic pollutants, pharmaceuticals and personal care products (PPCPs), and other organic contaminants in sludge from seven Swedish sewage treatment plants (STPs) was performed in this study. This extensive screening provides information on mass flows of
Timothy Lynagh et al.
The Journal of biological chemistry, 286(51), 43913-43924 (2011-10-29)
Ivermectin is an anthelmintic drug that works by activating glutamate-gated chloride channel receptors (GluClRs) in nematode parasites. GluClRs belong to the Cys-loop receptor family that also includes glycine receptor (GlyR) chloride channels. GluClRs and A288G mutant GlyRs are both activated
Highly diastereoselective multicomponent cascade reactions: efficient synthesis of functionalized 1-indanols.
Jun Jiang et al.
Angewandte Chemie (International ed. in English), 52(5), 1539-1542 (2013-01-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service