Skip to Content
Merck
All Photos(1)

Key Documents

B38503

Sigma-Aldrich

Bis(2-chloroethyl)amine hydrochloride

98%

Synonym(s):

β,β′-Dichlorodiethylamine hydrochloride, 1,5-Dichloro-3-azapentane hydrochloride, 2,2′-Dichlorodiethylamine hydrochloride, 2-Chloro-N-(2-chloroethyl)ethanamine hydrochloride, Bi(2-chloroethyl)amine hydrochloride, Bis(β-chloroethyl)amine hydrochloride, Bis(2-chloroethyl)ammonium chloride, Di(2-chloroethyl)amine hydrochloride, N,N-Bis(2-chloroethyl)amine hydrochloride, Nornitrogen mustard hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCH2CH2)2NH·HCl
CAS Number:
Molecular Weight:
178.49
Beilstein:
3550356
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

212-214 °C (lit.)

SMILES string

Cl.ClCCNCCCl

InChI

1S/C4H9Cl2N.ClH/c5-1-3-7-4-2-6;/h7H,1-4H2;1H

InChI key

YMDZDFSUDFLGMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-chloroethyl)amine hydrochloride is used as a starting material to synthesize piperazine derivatives.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

No information available.

Flash Point(C)

No information available.

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Cytarska et al.
Saudi pharmaceutical journal : SPJ : the official publication of the Saudi Pharmaceutical Society, 27(3), 303-311 (2019-04-13)
Synthesis, characterization and investigation of antiproliferative activity of nine triazene salts against human cancer cells lines (MV-4-11, MCF-7, JURKAT, HT-29, Hep-G2, HeLa, Du-145 and DAUDI), and normal human mammary epithelial cell line (MCF7-10A) is presented. The structures of novel compounds
R Fleer et al.
Chemico-biological interactions, 39(1), 1-15 (1982-03-01)
Activated cyclophosphamide (CP) is known to achieve its cytotoxic and alkylating capacity upon spontaneous hydrolytic breakdown of the oxazaphosphorine ring structure. Treatment of yeast cells with the chemically activated form of CP (4-hydroperoxy-CP, 4-OOH-CP) and with several potentially toxic cleavage
Daan Noort et al.
Archives of toxicology, 76(2), 83-88 (2002-03-27)
Covalent binding of various clinically important nitrogen mustards to the cysteine-34 residue of human serum albumin, in vitro and in vivo, is demonstrated. A rapid method for detection of these adducts is presented, based on liquid chromatography-tandem mass spectrometry analysis
H Thulin et al.
Toxicology and industrial health, 11(1), 89-97 (1995-01-01)
Nor-nitrogen mustard is an alkylating agent used in kilogram quantities in the production of various cytostatics. It has been shown to be mutagenic in vitro, and therefore must be regarded as a health hazard. We have studied the environment and
B Hartley-Asp
The Prostate, 5(1), 93-100 (1984-01-01)
In growth proliferation experiments on two human prostatic carcinoma cell lines, DU 145 cells were found to be more sensitive to the cytotoxic effect of estramustine and nor-nitrogen mustard than PC-3 cells. Estramustine was, however, much more cytotoxic in both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service