Skip to Content
Merck
All Photos(1)

Key Documents

C0253

Sigma-Aldrich

Chlorambucil

Synonym(s):

4-(4-[Bis(2-chloroethyl)amino]phenyl)butyric acid, 4-[Bis(2-chloroethyl)amino]benzenebutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19Cl2NO2
CAS Number:
Molecular Weight:
304.21
Beilstein:
999011
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

OC(=O)CCCc1ccc(cc1)N(CCCl)CCCl

InChI

1S/C14H19Cl2NO2/c15-8-10-17(11-9-16)13-6-4-12(5-7-13)2-1-3-14(18)19/h4-7H,1-3,8-11H2,(H,18,19)

InChI key

JCKYGMPEJWAADB-UHFFFAOYSA-N

Gene Information

human ... CYP2D6(1565)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Chlorambucil has been used:
  • as an alkylating/chemotherapeutic agent to study its anti-inflammatory effects on amyloidogenesis in mice
  • as a positive control to determine the cytotoxicity in murine leukemic monocyte macrophages
  • as an adenine-specific DNA-alkylating reagent in the construction of multifunctional conjugate (8950A-Chb(Cl/OH) for mutation-specific DNA alkylation in HeLa S3 cells

Biochem/physiol Actions

Chlorambucil is a nitrogen mustard alkylating agent that acts as an anti-cancer drug. Chlorambucil binds to membranes, RNA, DNA, and proteins. It exhibits anti-inflammatory and anti-tumor activity by DNA cross-linking. Chlorambucil shows therapeutic effects against chronic lymphocytic leukemia (CLL) via a p53-dependent mechanism.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shrabani Barman et al.
Angewandte Chemie (International ed. in English), 55(13), 4194-4198 (2016-02-27)
Among the well-known phototriggers, the p-hydroxyphenacyl (pHP) group has consistently enabled the very fast, efficient, and high-conversion release of active molecules. Despite this unique behavior, the pHP group has been ignored as a delivery agent, particularly in the area of
C Muller et al.
Blood, 92(7), 2213-2219 (1998-09-25)
The objective of this study is to investigate the role of DNA-dependent protein kinase (DNA-PK) in the chronic lymphocytic leukemia (CLL) lymphocyte response to nitrogen mustard therapy. DNA-PK is a nuclear serine/threonine kinase that functions in DNA double-strand break repair
Takuya Hidaka et al.
Cell chemical biology, 29(4), 690-695 (2021-08-28)
Mutations in mitochondrial DNA (mtDNA) cause mitochondrial diseases, characterized by abnormal mitochondrial function. Although eliminating mutated mtDNA has potential to cure mitochondrial diseases, no chemical-based drugs in clinical trials are capable of selective modulation of mtDNA mutations. Here, we construct
Palle J Pedersen et al.
Journal of medicinal chemistry, 52(10), 3408-3415 (2009-05-01)
The synthesis and biophysical characterization of four prodrug ether phospholipid conjugates are described. The lipids are prepared from the anticancer drug chlorambucil and have C16 and C18 ether chains with phosphatidylcholine or phosphatidylglycerol headgroups. All four prodrugs have the ability
Shmuel Shtrasburg et al.
Autoimmunity reviews, 12(1), 18-21 (2012-08-11)
The mainstay of AA amyloidosis prevention and treatment is suppression of inflammation. In the present study we have tried to determine the efficacy of a variety of anti-inflammatory agents at suppressing AA amyloidosis in a mouse model of the disease.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service