Skip to Content
Merck
All Photos(1)

Key Documents

B19800

Sigma-Aldrich

S-Benzyl-L-cysteine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2SCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
211.28
Beilstein:
1879358
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

optical activity

[α]20/D +23°, c = 2 in 1 M NaOH

reaction suitability

reaction type: solution phase peptide synthesis

color

white to off-white

mp

214 °C (dec.) (lit.)

application(s)

detection
peptide synthesis

SMILES string

N[C@@H](CSCc1ccccc1)C(O)=O

InChI

1S/C10H13NO2S/c11-9(10(12)13)7-14-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)/t9-/m0/s1

InChI key

GHBAYRBVXCRIHT-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Michelle Rudden et al.
Scientific reports, 10(1), 12500-12500 (2020-07-29)
Body odour is a characteristic trait of Homo sapiens, however its role in human behaviour and evolution is poorly understood. Remarkably, body odour is linked to the presence of a few species of commensal microbes. Herein we discover a bacterial
M Inoue et al.
Biochemical pharmacology, 36(13), 2145-2150 (1987-07-01)
Biosynthesis of N-acetylcysteine S-conjugates of toxic electrophiles, mercapturic acids, occurs via inter-organ metabolism and transport in which liver, small intestine and kidney play an important role. Since a mercapturic acid is a hydrophobic organic anion and strongly binds to plasma
B E Hall et al.
Xenobiotica; the fate of foreign compounds in biological systems, 16(7), 609-614 (1986-07-01)
Benzylmercapturic acid is a major metabolite of [methylene-14C]benzyl chloride in the marmoset, as in the rat. The excretion of the minor metabolites benzylmercapturic acid sulphoxide and benzylcysteine accounted for a greater proportion of the dose than in the rat. Excretion
Mercapturic acid formation in cultured opossum kidney cells.
N Golenhofen et al.
Toxicology letters, 53(1-2), 261-261 (1990-09-01)
A E Pegg et al.
Biochemistry, 32(45), 11998-12006 (1993-11-16)
Human O6-alkylguanine-DNA alkyltransferase was rapidly inactivated by low concentrations of O6-benzylguanine, but the alkyltransferase from the Escherichia coli ogt gene was much less sensitive and alkyltransferases from the E. coli ada gene or from yeast were not affected. O6-Benzyl-2'-deoxyguanosine was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service