B11907
1-Phenyl-1,3-butanedione
99%
Synonym(s):
Benzoylacetone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5COCH2COCH3
CAS Number:
Molecular Weight:
162.19
Beilstein:
742413
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
powder
mp
54-56 °C (lit.)
density
1.09 g/mL at 25 °C (lit.)
SMILES string
CC(=O)CC(=O)c1ccccc1
InChI
1S/C10H10O2/c1-8(11)7-10(12)9-5-3-2-4-6-9/h2-6H,7H2,1H3
InChI key
CVBUKMMMRLOKQR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yasuko Yamada Maruo et al.
Analytica chimica acta, 702(2), 247-253 (2011-08-16)
We have developed a portable device for formaldehyde monitoring with both high sensitivity and high temporal resolution, and carried out indoor air formaldehyde concentration analysis. The absorbance difference of the sensor element was measured in the monitoring device at regular
Arpad Mihai Rostas et al.
Molecules (Basel, Switzerland), 25(17) (2020-08-23)
Complexes with mixed ligands [Cu(N-N)2(pmtp)](ClO4)2 ((1) N-N: 2,2'-bipyridine; (2) L: 1,10-phenanthroline and pmpt: 5-phenyl-7-methyl-1,2,4-triazolo[1,5-a]pyrimidine) were synthesized and structurally and biologically characterized. Compound (1) crystallizes into space group Pa and (2) in P-1. Both complexes display an intermediate stereochemistry between the
Linbo Xia et al.
Analytical chemistry, 76(10), 2910-2915 (2004-05-18)
A new method of single-drop microextraction combined with low-temperature electrothermal vaporization (LTETV)-ICPMS was proposed for the determination of trace Be, Co, Pd, and Cd with benzoylacetone (BZA) as both extractant and chemical modifier. Several factors that influence the microextraction efficiency
Ahmed Ahmed Omran
Chemical & pharmaceutical bulletin, 53(11), 1498-1501 (2005-11-08)
A rapid, sensitive and selective spectrophotometric method has been developed for the quantitative determination of dapsone (DAP) and metoclopramide hydrochloride (MCP) in both pure and dosage forms. Individual and simultaneous methods are based on the diazo coupling reaction of these
S F Tayyari et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(2), 394-404 (2006-07-18)
Molecular structure and vibrational frequencies of 1,3-diphenyl-1,3-propanedione, known as dibenzoylmethane (DBM), have been investigated by means of density functional theory (DFT) calculations. The results were compared with those of benzoylacetone (BA) and acetylacetone (AA), the parent molecule. IR and Raman
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service