Skip to Content
Merck
All Photos(1)

Documents

217042

Sigma-Aldrich

4,4,4-Trifluoro-1-phenyl-1,3-butanedione

99%

Synonym(s):

BTA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2COCF3
CAS Number:
Molecular Weight:
216.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

224 °C (lit.)

mp

38-40 °C (lit.)

solubility

95% ethanol: soluble 25 mg/mL, clear, colorless to yellow

SMILES string

FC(F)(F)C(=O)CC(=O)c1ccccc1

InChI

1S/C10H7F3O2/c11-10(12,13)9(15)6-8(14)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

VVXLFFIFNVKFBD-UHFFFAOYSA-N

Application

4,4,4-Trifluoro-1-phenyl-1,3-butanedione was used in the synthesis of series of NNO ketoimines bearing trifluoromethyl substituents via Schiff base condensation reaction. It was also used in mixed-ligand chelate extraction of trivalent lanthanides and as a ligand in the preparation of ternary lanthanide (Ln) complexes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses, crystal structures, visible and near-IR luminescent properties of ternary lanthanide (Dy3+, Tm3+) complexes containing 4, 4, 4-trifluoro-1-phenyl-1, 3-butanedione and 1, 10-phenanthroline.
Feng J, et al.
Journal of Luminescence, 128(12), 1957-1964 (2008)
Meixia Shan et al.
Science advances, 4(9), eaau1698-eaau1698 (2018-09-27)
The development of new membranes with high H2 separation performance under industrially relevant conditions (high temperatures and pressures) is of primary importance. For instance, these membranes may facilitate the implementation of energy-efficient precombustion CO2 capture or reduce energy intensity in
M E Grobman et al.
Journal of veterinary internal medicine, 33(3), 1423-1433 (2019-04-11)
Megaesophagus (ME) carries a poor long-term prognosis in dogs. In people, lower esophageal sphincter (LES) achalasia is a rare cause of ME that may respond to targeted intervention. Dogs with lower esophageal sphincter achalasia-like syndrome (LES-AS) have been described recently
Jean-Jacques Body et al.
Journal of bone oncology, 14, 100212-100212 (2019-01-11)
Bone metastases (BMs) are common in patients with prostate cancer and can lead to skeletal-related events (SREs), which are associated with increased pain and reduced quality of life (QoL). Bone-targeted agents (BTAs), such as zoledronic acid and denosumab, reduce the
Catarina Xavier et al.
Forensic science international. Genetics, 48, 102336-102336 (2020-07-04)
Forensic DNA phenotyping is gaining interest as the number of applications increases within the forensic genetics community. The possibility of providing investigative leads in addition to conventional DNA profiling for human identification provides new insights into otherwise "cold" police investigations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service