Recommended Products
Quality Level
Assay
95%
form
liquid
reaction suitability
reaction type: click chemistry
refractive index
n/D 1.476
density
1.421 at 25 °C
SMILES string
FC1=CC=CC=C1S(=O)(F)=O
InChI
1S/C6H4F2O2S/c7-5-3-1-2-4-6(5)11(8,9)10/h1-4H
InChI key
ODVXTZVBPYPQLJ-UHFFFAOYSA-N
Application
Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
224.0 °F
Flash Point(C)
106.67 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service