Skip to Content
Merck
All Photos(1)

Documents

108138

Sigma-Aldrich

Benzenesulfonyl chloride

99%

Synonym(s):

Phenylsulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2Cl
CAS Number:
Molecular Weight:
176.62
Beilstein:
606926
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39092406
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.04 mmHg ( 20 °C)

Quality Level

Assay

99%

refractive index

n20/D 1.551 (lit.)

bp

251-252 °C (lit.)

mp

13-15 °C (lit.)

solubility

alcohol: soluble
cold water: insoluble
diethyl ether: soluble

density

1.384 g/mL at 25 °C (lit.)

SMILES string

ClS(=O)(=O)c1ccccc1

InChI

1S/C6H5ClO2S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H

InChI key

CSKNSYBAZOQPLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzenesulfonyl chloride is prepared by reaction of benzene and chlorosulfonic acid or from the sodium salt of benzenesulfonic acid and PCl5 or POCl3. It reacts with Grignard reagent from N-unsubstituted indoles to form oxindoles or substituted indoles. Benzenesulfonyl chloride is the derivatization reagent for the determination of various amines in waste water and surface water at the sub-ppb level by gas chromatography-mass spectrometry.

Application

Benzenesulfonyl chloride may be used to develop fast, accurate and reproducible method for thiamine assay in different food products. It is useful reagent for preparing α-disulfones.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

260.6 °F

Flash Point(C)

127 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzenesulfonyl chloride
Adams R, et al.
Organic Syntheses, 1(84) (1921)
Synthesis, 490-490 (1993)
A G Soliman
Journal - Association of Official Analytical Chemists, 64(3), 616-622 (1981-05-01)
A semiautomated procedure was used to measure the fluorescence of sample extracts before and after the addition of benzenesulfonyl chloride (BSC). Addition of BSC inhibited thiochrome formation and provided a more representative blank based on the fluorescence of all the
Zongren Li et al.
Chemistry, an Asian journal, 14(13), 2302-2308 (2019-05-12)
Here, we designed several waterborne polyurethanes (WPUs) with efficient thermally activated delayed fluorescence (TADF) via serving charge-transfer (CT) states as a mediate bridge between singlet and triplet states to boost reverse intersystem crossing (RISC). By tuning substituents of diphenyl sulfone
Analysis of primary and secondary aliphatic amines in waste water and surface water by gas chromatography-mass spectrometry after derivatization with 2, 4-dinitrofluorobenzene or benzenesulfonyl chloride.
Sacher F, et al.
Journal of Chromatography A, 764(1), 85-93 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service