ALD00110
Diethyl 1,2,3-triazine-4,6-dicarboxylate
≥95%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥95%
form
solid
storage temp.
2-8°C
SMILES string
O=C(OCC)C1=NN=NC(C(OCC)=O)=C1
InChI
1S/C9H11N3O4/c1-3-15-8(13)6-5-7(11-12-10-6)9(14)16-4-2/h5H,3-4H2,1-2H3
InChI key
WSXMBZCJYIARRQ-UHFFFAOYSA-N
Related Categories
Application
1,2,3-Triazines have been shown to be reactive substrates in inverse electron demand Diels-Alder strategies. Recent examples by the Boger Research Group have utilized this reactive motif in the construction of highly functionalized N-containing heterocycles.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service