Skip to Content
Merck
All Photos(1)

Key Documents

ALD00554

Sigma-Aldrich

5-phenyl-1,2,3-triazine

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N3
CAS Number:
Molecular Weight:
157.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

flakes

mp

141-146 °C

storage temp.

−20°C

SMILES string

C1(C2=CC=CC=C2)=CN=NN=C1

InChI

1S/C9H7N3/c1-2-4-8(5-3-1)9-6-10-12-11-7-9/h1-7H

InChI key

KJZQIXWSZPPOHO-UHFFFAOYSA-N

General description

5-Phenyl-1,2,3-triazine is a phenyl triazine derivative. 5-phenyl-1,2,3-triazine exhibits electronic and nonlinear optical properties. 5-Phenyl-1,2,3-triazine can be prepared from 4-bromopyrazole. It undergoes Diels-Alder reaction with ketene acetal.

Application

The following 1,2,3-triazine was reported by Boger and coworkers to undergo an Inverse Electron Demand Diels-Alder with electron rich dienophiles to afford nitrogen-containing heterocycles, more specifically pyrimidines and novel-substituted pyridines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of torsional barriers and nonlinear optical (NLO) properties of phenyltriazines.
Alyar H, et al.
Journal of Molecular Structure, 834, 516-520 (2007)
Erin D Anderson et al.
Journal of the American Chemical Society, 133(31), 12285-12292 (2011-07-09)
A systematic study of the inverse electron demand Diels-Alder reactions of 1,2,3-triazines is disclosed, including an examination of the impact of a C5 substituent. Such substituents were found to exhibit a remarkable impact on the cycloaddition reactivity of the 1,2,3-triazine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service