Skip to Content
Merck
All Photos(1)

Documents

A56655

Sigma-Aldrich

4-Amino-3-hydroxybutyric acid

98%

Synonym(s):

DL-γ-Amino-β-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH(OH)CH2CO2H
CAS Number:
Molecular Weight:
119.12
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white to yellow

mp

223 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NCC(O)CC(O)=O

InChI

1S/C4H9NO3/c5-2-3(6)1-4(7)8/h3,6H,1-2,5H2,(H,7,8)

InChI key

YQGDEPYYFWUPGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T W Stone
European journal of pharmacology, 128(1-2), 81-83 (1986-08-22)
Kynurenine and kynurenic acid are known to produce convulsions in rats and mice and it has been reported that kynurenine can displace GABA from its neuronal binding sites. The present study shows that neither kynurenine nor kynurenic acid are antagonist
D Jordan et al.
Brain research, 268(1), 105-110 (1983-05-23)
Various doses of GABA from 0.25 to 5 mumol injected into the third ventricle decrease serum TSH rapidly. The same effect was observed with GABOB (10 mumol), the hydroxylated form of GABA. The inhibitory effect of both of these drugs
M Candela et al.
Journal of chromatography. A, 890(2), 273-280 (2000-09-29)
A rapid and simple reversed-phase liquid chromatographic method that did not require the derivatization of 4-amino-3-hydroxybutyric acid (GABOB) was developed and validated. The method proved to be suitable for the determination of GABOB concentrations in finished pharmaceutical product (tablets). The
[Neurochemical aspects of the pharmacology of GABAergic substances].
K S Raevskiĭ
Farmakologiia i toksikologiia, 44(5), 517-529 (1981-09-01)
Izumi Yamamoto et al.
ACS chemical neuroscience, 3(9), 665-673 (2012-09-29)
Designing potent and subtype-selective ligands with therapeutic value requires knowledge about how endogenous ligands interact with their binding site. 4-Amino-3-hydroxybutanoic acid (GABOB) is an endogenous ligand found in the central nervous system in mammals. It is a metabolic product of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service