Skip to Content
Merck
All Photos(1)

Key Documents

A48407

Sigma-Aldrich

4-Amino-2,6-dichlorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H2(Cl)2OH
CAS Number:
Molecular Weight:
178.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

167-170 °C (lit.)

SMILES string

Nc1cc(Cl)c(O)c(Cl)c1

InChI

1S/C6H5Cl2NO/c7-4-1-3(9)2-5(8)6(4)10/h1-2,10H,9H2

InChI key

KGEXISHTCZHGFT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Aigner et al.
Journal of materials chemistry. C, 1(36), 5685-5693 (2013-10-01)
New optical pH-sensors relying on 1,4-diketopyrrolo-[3,4-
Benoît Limoges et al.
Journal of the American Chemical Society, 130(23), 7259-7275 (2008-05-21)
The use of enzyme labeling techniques to convert biorecognition events into high sensitivity electrochemical signals may follow two different strategies. One, in which the current is the electrocatalytic response of a redox couple serving as cosubstrate to a redox enzyme
Gary O Rankin et al.
Toxicology, 245(1-2), 123-129 (2008-02-05)
4-Amino-2,6-dichlorophenol (ADCP) is a potent acute nephrotoxicant in vivo inducing prominent renal corticomedullary necrosis. In vitro, ADCP exposure increases lactate dehydrogenase (LDH) release from rat renal cortical slices at 0.05 mM or greater. The purpose of this study was to
Benoît Limoges et al.
Journal of the American Chemical Society, 130(23), 7276-7285 (2008-05-22)
The two articles in this series are dedicated to bioaffinity electrodes with in situ detection of the product of the enzyme label after recognition by its conjugate immobilized on the electrode. Part 1 was devoted to direct electrochemical detection, whereas
S K Hong et al.
Toxicology and applied pharmacology, 147(1), 115-125 (1997-11-14)
A halogenated derivative of 4-aminophenol, 4-amino-2, 6-dichlorophenol (ADCP), is a potent nephrotoxicant and a weak hepatotoxicant in Fischer 344 rats. Although the mechanism of ADCP nephrotoxicity is unknown, ADCP could undergo oxidation to a reactive intermediate, such as a 4-amino-2,6-dichlorophenoxy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service